Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Curved, 105 mm
Catalog # 232-0008
Supplier:   BOCHEM
Forceps, precision
Catalog # 232-0008
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Curved, 105 mm
Price
239,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production and more.
  • Extremely sharp, flat
  • Non serrated points
Specifications
  • Type:
    Curved
  • Délka:
    105 mm