Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
01, 19/26 NS, 19/26 NS
  201-8572
 :   SciLabware
Multiple adapters, Quickfit®
  201-8572
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
01, 19/26 NS, 19/26 NS
2 340,00 Kč
1 * 1 KS
PYREX®, borosilicate glass, straight.
  • Both sockets are the same size
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters with two parallel necks/sockets and a cone.
  • Rozměry zábrusu:
    19/26 NS
  • Velikost objímky:
    19/26 NS
  • Sekvence:
    01