Orders LinkContactUsLinkComponent
Separating funnels, Gilson
Separating funnels, Gilson
11, 500 ml, 24/29 NS
Katalogové číslo BIBB443054N
Dodavatel:   SciLabware
Separating funnels, Gilson
Katalogové číslo BIBB443054N
Dodavatel:  SciLabware
Separating funnels, Gilson
Separating funnels, Gilson
11, 500 ml, 24/29 NS
Přejít na:
Price
4 200,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
PYREX®, borosilicate glass, with NS-PE stopper.
Gilson style separating funnel with long delivery stem for good control of the liquid/liquid interface. With interchangeable glass or PTFE key for grease free performance. Supplied with hollow glass stopper.
Specifikace
  • Rozměry zábrusu:
    24/29 NS
  • Kapacita:
    500 ml
  • Sekvence:
    11