Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Replacement blade, stainless steel, medium, 31,75×14,29 mm
Katalogové číslo BELC7731-22115
Marketsource
Dodavatel:   BELLCO GLASS
Cell scrapers
Katalogové číslo BELC7731-22115
Dodavatel:  BELLCO GLASS
Marketsource
Cell scrapers
Cell scrapers
Replacement blade, stainless steel, medium, 31,75×14,29 mm
Přejít na:
Price
3 270,00 Kč
Přihlásit se pro dostupnost
1 * 10 KS
Product Details & Documents
Stainless steel blade holder mounted on plastic covered stainless steel handle.
Specifikace
  • Popis:
    Replacement blade, stainless steel, medium, 31,75×14,29 mm