Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Micro capillaries, disposable, minicaps

Micro capillaries, disposable, minicaps

03, Sodium-heparinised minicaps, 3 µl, 30 - 32 mm
Catalog # 720-2836
Supplier:   HIRSCHMANN
Micro capillaries, disposable, minicaps
Catalog # 720-2836
Supplier:  HIRSCHMANN
Micro capillaries, disposable, minicaps
Micro capillaries, disposable, minicaps

03, Sodium-heparinised minicaps, 3 µl, 30 - 32 mm

Price
2 920,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents

Disposable capillary pipettes with the end-to-end system. With genuine Schott DURAN® glass. Accuracy ±≤0,5%, CV ±≤1%. With batch number on the package.

  • Correct filling of the capillary
  • Conformity certified, according to the German 'Eichordnung' certification
  • Colour-coded vial cap with capillary dispenser
  • Safe packaging for protection during transportation

Packaging: 100 minicaps packed in a glass vial with dispenser cap. 10 vials plus a pipetting device are packed into a outer carton.

Specifications
  • Popis:
    Sodium-heparinised minicaps, 3 µl
  • Délka:
    30 - 32 mm
  • Sekvence:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.