Orders LinkContactUsLinkComponent
Volumetric flasks, class B
1000 ml, 24/29, 310 mm
  HIRS7550193
 :   HIRSCHMANN
Volumetric flasks, class B
  HIRS7550193
Volumetric flasks, class B
Volumetric flasks, class B
1000 ml, 24/29, 310 mm
605,00 Kč
1 * 1 KS
PP, translucent, with PP NS-stopper.
  • NZ:
    24/29
  • Kapacita:
    1000 ml
  • Výška:
    310 mm