Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Handle, stainless steel, for staining rack 631-9321, - mm
Catalog # 631-9329
Staining jars
Catalog # 631-9329
Staining jars
Staining jars
Handle, stainless steel, for staining rack 631-9321, - mm
Price
108,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Soda-lime glass.
  • For 10 standard slides  (76×26 mm)
Staining system contains staining jar, rack and handle made from stainless steel.
Ordering information: Insert and handle must be ordered separately.
Specifications
  • Popis:
    Handle, stainless steel, for staining rack 631-9321
  • Š×H×V:
    - mm