Staining jars
Staining dishes acc. to Schiefferdecker, for up to 10 slides
Katalogové číslo 631-9313
Dodavatel: GLASWARENFABRIK KARL HECHT
Přejít na:
Product Details & Documents
Soda-lime glass.
- Three versions
- For 16 or 10 slides
- Acc. to Hellendahl or Schiefferdecker
Suitable for standard microscope slides (76×26 mm).
Specifikace
- Popis:Staining dishes acc. to Schiefferdecker, for up to 10 slides