Orders LinkContactUsLinkComponent
Staining jars
Staining jars
Rectangular staining jar with cover, 40×20×85 mm, - mm, - mm
Katalogové číslo 631-9314
Staining jars
Katalogové číslo 631-9314
Staining jars
Staining jars
Rectangular staining jar with cover, 40×20×85 mm, - mm, - mm
Přejít na:
Price
288,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Soda-lime glass.
These staining jars have an overlapping cover.
Specifikace
  • Ø:
    - mm
  • Popis:
    Rectangular staining jar with cover
  • Výška:
    - mm
  • Š×H×V:
    40×20×85 mm