Orders LinkContactUsLinkComponent
Staining dishes
Staining dish with lid, without tray, 90×70×65 mm
  631-0347
Staining dishes
  631-0347
Staining dishes
Staining dishes
Staining dish with lid, without tray, 90×70×65 mm
771,00 Kč
1 * 1 KS
Soda-lime glass.
  • Removable cover
Staining dish for 76×26 mm slides or slides with width up to 45 mm.
  • Popis:
    Staining dish with lid, without tray
  • Š×H×V:
    90×70×65 mm
Accessories
  •  
    Popis
    Š×H×V
  • 631-0864
    Lid for staining dish
    - mm
    1 * 1 KS - 288,00 Kč
  •  
    Popis
    Š×H×V
  • 631-0348
    Stainless steel rack for staining jars, for 10 slides
    - mm
    1 * 1 KS - 495,00 Kč