Orders LinkContactUsLinkComponent
Forceps, straight
Forceps, straight
Sharp tips, 120 mm
Catalog # 232-2151
Supplier:   HAMMACHER KARL
Forceps, straight
Catalog # 232-2151
Supplier:  HAMMACHER KARL
Forceps, straight
Forceps, straight
Sharp tips, 120 mm
Price
1 290,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic.
  • Smooth points
Specifications
  • Type:
    Sharp tips
  • Délka:
    120 mm