Orders LinkContactUsLinkComponent
Light forceps
Light forceps
01, Blunt, 100 mm
Catalog # 232-2120
Supplier:   HAMMACHER KARL
Light forceps
Catalog # 232-2120
Supplier:  HAMMACHER KARL
Light forceps
Light forceps
01, Blunt, 100 mm
Price
1 290,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Light forceps made of watch-spring sheet-steel.
Specifications
  • Type:
    Blunt
  • Délka:
    100 mm
  • Sekvence:
    01