Physiology scissors, straight
Sharp/sharp, 145 mm
Katalogové číslo 233-0086
Dodavatel: HAMMACHER KARL
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- Sharp-sharp
- Ideal for a wide range of laboratory applications
- Acid-resistant
- Autoclavable
Specifikace
- Type:Sharp/sharp
- Délka:145 mm