Thread and vessel scissors
Straight, sharp/blunt, 130 mm
Katalogové číslo 233-0083
Dodavatel: HAMMACHER KARL
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- Sharp-blunt
- Ideal for a wide range of laboratory applications
- Acid-resistant
- Autoclavable
Specifikace
- Type:Straight, sharp/blunt
- Délka:130 mm