Orders LinkContactUsLinkComponent
Dissecting scissors
Dissecting scissors
Curved, sharp/sharp, 110 mm
Catalog # HAMMHWB005-11
Supplier:   HAMMACHER KARL
Dissecting scissors
Catalog # HAMMHWB005-11
Supplier:  HAMMACHER KARL
Dissecting scissors
Dissecting scissors
Curved, sharp/sharp, 110 mm
Price
3 200,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • Ideal for a wide range of laboratory applications
  • Acid-resistant
  • Autoclavable
  • Sharp-sharp
Specifications
  • Type:
    Curved, sharp/sharp
  • Délka:
    110 mm