Dissecting scissors
Straight, sharp/sharp, 110 mm
Katalogové číslo 233-1104
Dodavatel: HAMMACHER KARL
Přejít na:
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
- Ideal for a wide range of laboratory applications
- Acid-resistant
- Autoclavable
- Sharp-sharp
Specifikace
- Type:Straight, sharp/sharp
- Délka:110 mm