Orders LinkContactUsLinkComponent
Poly-spoons
Poly-spoons
07, One broad spoon end, one spatula end, 300 mm
Catalog # HAMMHSN175-30
Supplier:   HAMMACHER KARL
Poly-spoons
Catalog # HAMMHSN175-30
Supplier:  HAMMACHER KARL
Poly-spoons
Poly-spoons
07, One broad spoon end, one spatula end, 300 mm
Price
794,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Poly-spoons made of stainless-steel.
  • Double-ended: One spoon and one spatula end
Specifications
  • Popis:
    One broad spoon end, one spatula end
  • Délka:
    300 mm
  • Sekvence:
    07