Orders LinkContactUsLinkComponent
Micro forceps
Micro forceps
01, No. 3, straight, very fine tips, 115 mm
Catalog # HAMMHSC600-11
Supplier:   HAMMACHER KARL
Micro forceps
Catalog # HAMMHSC600-11
Supplier:  HAMMACHER KARL
Micro forceps
Micro forceps
01, No. 3, straight, very fine tips, 115 mm
Price
1 670,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    No. 3, straight, very fine tips
  • Délka:
    115 mm
  • Sekvence:
    01