Orders LinkContactUsLinkComponent
Microscopic forceps
Microscopic forceps
12, Microscopic forceps, Strongly curved, 145 mm
Catalog # HAMMHSC565-14
Supplier:   HAMMACHER KARL
Microscopic forceps
Catalog # HAMMHSC565-14
Supplier:  HAMMACHER KARL
Microscopic forceps
Microscopic forceps
12, Microscopic forceps, Strongly curved, 145 mm
Price
1 500,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Type:
    Strongly curved
  • Popis:
    Microscopic forceps
  • Délka:
    145 mm
  • Sekvence:
    12