Orders LinkContactUsLinkComponent
Microscopic forceps
Microscopic forceps
01, Microscopic forceps, Straight, 105 mm
Catalog # HAMMHSC550-10
Supplier:   HAMMACHER KARL
Microscopic forceps
Catalog # HAMMHSC550-10
Supplier:  HAMMACHER KARL
Microscopic forceps
Microscopic forceps
01, Microscopic forceps, Straight, 105 mm
Price
1 290,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Specifications
  • Type:
    Straight
  • Popis:
    Microscopic forceps
  • Délka:
    105 mm
  • Sekvence:
    01