Orders LinkContactUsLinkComponent
Preparation forceps
Preparation forceps
05, Straight, fine, 115 mm
Catalog # HAMMHSC186-11
Supplier:   HAMMACHER KARL
Preparation forceps
Catalog # HAMMHSC186-11
Supplier:  HAMMACHER KARL
Preparation forceps
Preparation forceps
05, Straight, fine, 115 mm
Price
939,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, fine
  • Délka:
    115 mm
  • Sekvence:
    05