Orders LinkContactUsLinkComponent
Specimen seizing forceps
Specimen seizing forceps
01, Specimen seizing forceps, Taylor, Straight, fine tips, 180 mm
Catalog # 232-2103
Supplier:   HAMMACHER KARL
Specimen seizing forceps
Catalog # 232-2103
Supplier:  HAMMACHER KARL
Specimen seizing forceps
Specimen seizing forceps
01, Specimen seizing forceps, Taylor, Straight, fine tips, 180 mm
Price
2 060,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Stainless steel.
Specifications
  • Type:
    Straight, fine tips
  • Popis:
    Specimen seizing forceps, Taylor
  • Délka:
    180 mm
  • Sekvence:
    01