Orders LinkContactUsLinkComponent
Dressing forceps, delicate
Dressing forceps, delicate
03, Delicate model, Straight, blunt, 145 mm
Catalog # HAMMHSC074-14
Supplier:   HAMMACHER KARL
Dressing forceps, delicate
Catalog # HAMMHSC074-14
Supplier:  HAMMACHER KARL
Dressing forceps, delicate
Dressing forceps, delicate
03, Delicate model, Straight, blunt, 145 mm
Price
906,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Delicate dressing forceps made from stainless steel.
Specifications
  • Type:
    Straight, blunt
  • Popis:
    Delicate model
  • Délka:
    145 mm
  • Sekvence:
    03