Orders LinkContactUsLinkComponent
Dissecting scissors
Dissecting scissors
01, Straight, sharp/sharp, 115 mm
Catalog # HAMML-HSB300-11
Supplier:   HAMMACHER KARL
Dissecting scissors
Catalog # HAMML-HSB300-11
Supplier:  HAMMACHER KARL
Dissecting scissors
Dissecting scissors
01, Straight, sharp/sharp, 115 mm
Price
7 010,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These dissecting scissors made of stainless steel feature tungsten carbide inserts.
Specifications
  • Type:
    Straight, sharp/sharp
  • Délka:
    115 mm
  • Sekvence:
    01