Orders LinkContactUsLinkComponent
Dissecting scissors, type Joseph
Dissecting scissors, type Joseph
01, Joseph, straight, sharp/sharp, 140 mm
Katalogové číslo HAMML-HSB028-15
Dodavatel:   HAMMACHER KARL
Dissecting scissors, type Joseph
Katalogové číslo HAMML-HSB028-15
Dodavatel:  HAMMACHER KARL
Dissecting scissors, type Joseph
Dissecting scissors, type Joseph
01, Joseph, straight, sharp/sharp, 140 mm
Price
2 280,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
Dissecting scissors made of stainless steel.
Documents
Specifications
  • Type:
    Joseph, straight, sharp/sharp
  • Délka:
    140 mm
  • Sekvence:
    01