Orders LinkContactUsLinkComponent
Microscopy scissors
Microscopy scissors
02, Curved, sharp/sharp, with angular handles, 105 mm
Catalog # HAMML-HSB011-10
Supplier:   HAMMACHER KARL
Microscopy scissors
Catalog # HAMML-HSB011-10
Supplier:  HAMMACHER KARL
Microscopy scissors
Microscopy scissors
02, Curved, sharp/sharp, with angular handles, 105 mm
Price
1 620,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These sharp microscopy scissors with angular handles are made from stainless steel.
Specifications
  • Type:
    Curved, sharp/sharp, with angular handles
  • Délka:
    105 mm
  • Sekvence:
    02