Büchner funnels
08, 580 ml, 110 mm
Katalogové číslo 511-2507
Dodavatel: MORGAN ADVANCED MATERIALS HALD
Přejít na:
Product Details & Documents
White glazed porcelain.
For filter paper or membrane filtration discs.
Certifications: DIN 12 905 (511-2502, 511-2501 and 511-2500 not according to DIN).
Specifikace
- Kapacita:580 ml
- Sekvence:08
- Pro filtr Ø:110 mm