Orders LinkContactUsLinkComponent
Measuring spoons
Measuring spoons
02, Measuring spoon, molded plastic cup, black, 0,1 g
Katalogové číslo HACH51100
Dodavatel:   Hach
Measuring spoons
Katalogové číslo HACH51100
Dodavatel:  Hach
Measuring spoons
Measuring spoons
02, Measuring spoon, molded plastic cup, black, 0,1 g
Přejít na:
Price
227,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Molded plastic cup. Weight based on sodium chloride.
Specifikace
  • Kapacita:
    0,1 g
  • Popis:
    Measuring spoon, molded plastic cup, black
  • Sekvence:
    02