Orders LinkContactUsLinkComponent
Measuring spoons
Measuring spoons
05, Measuring spoon, molded plastic cup, black, 1,0 g
Katalogové číslo HACH51000
Dodavatel:   Hach
Measuring spoons
Katalogové číslo HACH51000
Dodavatel:  Hach
Measuring spoons
Measuring spoons
05, Measuring spoon, molded plastic cup, black, 1,0 g
Přejít na:
Price
168,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Molded plastic cup. Weight based on sodium chloride.
Specifikace
  • Kapacita:
    1,0 g
  • Popis:
    Measuring spoon, molded plastic cup, black
  • Sekvence:
    05