Microplates, 96-well
06, F-bottom, clear, sterile, 40 - 340 µl, 96, +, 382 µl
Catalog # 736-0119
Supplier: Greiner Bio-One
Jump to:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Documents
Specifications
- Poč. jamek:96
- Doporučený pracovní objem:40 - 340 µl
- Sterilní:+
- Popis:F-bottom, clear, sterile
- Sekvence:06
- Celkový objem:382 µl
- Cat. No.:736-0119
- Certifikát ochrany životního prostředí:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact