Microplates, 96-well
01, V-bottom, clear, 40 - 200 µl, 96, -, 234 µl
Dodavatel: Greiner Bio-One
Přejít na:
Product Details & Documents
PS, without lid. A large number of different 96-well microplates are available in a wide variety of surface treatments with a choice of well profile, and with either solid or µClear® bottom. All plates are free of DNase, RNase and human DNA, and are non-pyrogenic.
Specifikace
- Poč. jamek:96
- Doporučený pracovní objem:40 - 200 µl
- Sterilní:-
- Popis:V-bottom, clear
- Sekvence:01
- Celkový objem:234 µl
- Cat. No.:391-3610
- Certifikát ochrany životního prostředí:ISO 14001 (241002); ISO 50001 (241002)
- Environmentally Preferable:Low Manufacturing Impact