Automatic burettes, Schilling pattern
25 ml, 0,10 ml
Katalogové číslo POUL1.06843
Dodavatel: Poulten & Graf
Přejít na:
Product Details & Documents
AR-GLAS®, clear or amber, class B.
- Calibrated to deliver 'TD, Ex'
- Schellbach stripe, reduced distance between subdivision marks
- Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
Error limits according to class B, DIN EN ISO 385
Certifications: Error limits according to class B, DIN EN ISO 385
Specifikace
- Kapacita:25 ml
- Dělení:0,10 ml