Orders LinkContactUsLinkComponent
Temperature probes
Temperature probes
Temperature probe, with 1 m cable and water proof plug, TFK 325, Automatic temperature compensation in aqueous solutions
Katalogové číslo 662-0264
Dodavatel:   WTW
Temperature probes
Katalogové číslo 662-0264
Dodavatel:  WTW
Temperature probes
Temperature probes
Temperature probe, with 1 m cable and water proof plug, TFK 325, Automatic temperature compensation in aqueous solutions
Price
4 710,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Specifikace
  • Popis:
    Temperature probe, with 1 m cable and water proof plug
  • Typ:
    TFK 325
  • Aplikace:
    Automatic temperature compensation in aqueous solutions
Accessories
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • Holding clip for SenTix pH electrodes and temperature sensor TFK 325/HC
    Holding clip for SenTix pH electrodes and temperature sensor TFK 325/HC
    WTWA109821
    Přihlásit se pro dostupnost
    1 * 1 KS - 2 860,00 Kč