Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
[EN]BTEX STANDARD SOLUTION - 6 COMPONENT 1 * 1 mL

[EN]BTEX STANDARD SOLUTION - 6 COMPONENT 1 * 1 mL

Katalogové číslo ACSDF129231
New Product
Dodavatel:   CPACHEM
[EN]BTEX STANDARD SOLUTION - 6 COMPONENT 1 * 1 mL
Katalogové číslo ACSDF129231
Dodavatel:  CPACHEM
New Product
Číslo CAS:  
[EN]BTEX STANDARD SOLUTION - 6 COMPONENT 1 * 1 mL
Číslo CAS:  
Price
5 560,00 Kč
Přihlásit se pro dostupnost
1 * 1 mL
Product Details & Documents
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.