Volumetric Flasks, Class A
17, 15 ml, 10/19, 0,04 ± ml
Katalogové číslo WITG3.662.016.BL
New Product
Dodavatel: witeg Labortechnik
Volumetric Flasks, Class A
Katalogové číslo WITG3.662.016.BL
Dodavatel: witeg Labortechnik
New Product
Přejít na:
Product Details & Documents
Borosilicate glass 3.3, with or without stopper.
- DE-M marked according to the German calibration law
- Inclusive batch certificate
Certifications: Tolerance according to DIN EN ISO 1042
Specifikace
- NZ:10/19
- Kapacita:15 ml
- Sekvence:17
- Tolerance:0,04 ± ml