Digital Microscopes, Edge
1, Digital microscope, 1,3 MP with magnification lock, AF4115ZT, 20 ~ 220×, 8 white LEDs, 1280×960 pixels, Standard
Katalogové číslo IDCPAF4115ZT
New Product
Dodavatel: Dino-Lite / IDCP B.V.
Přejít na:
Product Details & Documents
The Dino-Lite Edge models provide superior image quality and a high level of flexibility due to the adaptable cap design. The built-in adjustable polarizer reduces glare and reflection on shiny objects.
This model can work with two magnification rates while maintaining the same working distance, making this the ideal model for macro view applications with a larger field of view and a zoom function to reveal smaller details. Different caps are supplied with the microscope making the Dino-Lite Edge the perfect choice for any professional application.
Specifikace
- Zvětšení:20 ~ 220×
- Model:AF4115ZT
- Rozlišení:1280×960 pixels
- Popis:Digital microscope, 1,3 MP with magnification lock
- Sekvence:1
- Osvětlení:8 white LEDs
- Pracovní vzdálenost:Standard
- Snímková frekvence:Up to 30 fps
- Hmotnost:111 g
- Cat. No.:IDCPAF4115ZT
- Požadavky na počítač:Windows XP, Vista, 7, 8, 10, Mac OS 10.11.6 or newer
- Kamera:Digital camera, 1,3 MP
- Snímač:CMOS
- Software:DinoCapture 2,0 (Windows), DinoXcope (Mac OS)