Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
NBD-Cl

NBD-Cl

3047, Bottle
Catalog # APOSBUP18438-1G
New Product
Supplier:   Apollo Scientific
NBD-Cl
Catalog # APOSBUP18438-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  
NBD-Cl

3047, Bottle

CAS Number:  
Price
1 440,00 Kč
Log in for availability
1 * 1 g
Product Details & Documents
Specifications

Specifications

  • Typ balení:
    Bottle
  • Sekvence:
    3047

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.