Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Robustine ≥98%

Robustine ≥98%

2370, Bottle
Catalog # APOSBUP00843-50MG
New Product
Supplier:   Apollo Scientific
Robustine ≥98%
Catalog # APOSBUP00843-50MG
Supplier:  Apollo Scientific
New Product
CAS Number:  2255-50-7
Robustine ≥98%

2370, Bottle

CAS Number:  2255-50-7
Price
28 370,00 Kč
Log in for availability
1 * 50 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Robustine
  • CAS Number:2255-50-7
  • Formula:C₁₂H₉NO₃
  • Molecular Weight:215,2 g/mol
Specifications

Specifications

  • Typ balení:
    Bottle
  • Sekvence:
    2370

Specifications