Ethionamide ≥98%
2725, Bottle
Katalogové číslo APOSBUP17752-500MG
New Product
Dodavatel: Apollo Scientific
Ethionamide ≥98%
Katalogové číslo APOSBUP17752-500MG
Dodavatel: Apollo Scientific
New Product
Číslo CAS: 536-33-4
Přejít na:
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyma: 2-Ethylpyridine-4-carbothioamide, 2-Ethylthioisonicotinamide
- Číslo CAS:536-33-4
- Vzorec:C₈H₁₀N₂S
- Molecular Weight:166,24 g/mol
Specifikace
Specifikace
- Typ balení:Bottle
- Sekvence:2725
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



