Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Doxycycline ≥95%

Doxycycline ≥95%

2725, Bottle
Catalog # APOSBUP17209-500MG
New Product
Supplier:   Apollo Scientific
Doxycycline ≥95%
Catalog # APOSBUP17209-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  564-25-0
Doxycycline ≥95%

2725, Bottle

CAS Number:  564-25-0
Price
6 280,00 Kč
Log in for availability
1 * 500 mg
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Vibramycin, Doxytetracycline, Doxiciclina
  • CAS Number:564-25-0
  • Formula:C₂₂H₂₄N₂O₈
  • Molecular Weight:444,43 g/mol
Specifications

Specifications

  • Typ balení:
    Bottle
  • Sekvence:
    2725

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.