Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Mitomycin

Mitomycin

2725, Bottle
Catalog # APOSBIM0133-500MG
Toxic
Health
New Product
Supplier:   Apollo Scientific
Mitomycin
Catalog # APOSBIM0133-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  50-07-7
Mitomycin

2725, Bottle

CAS Number:  50-07-7
Price
68 250,00 Kč
Log in for availability
1 * 500 mg
Product Details & Documents

Mitomycin C is a HIGHLY TOXIC antibiotic with antineoplastic properties.

Synonyms:  Ametycin
  • CAS Number:50-07-7
  • Formula:C₁₅H₁₈N₄O₅
  • Melting Pt:360 °C
  • Storage Temperature:Refrigerator
  • EINECS:200-008-6
  • UN:3462
  • ADR:6.1,II

Documents

Specifications

Specifications

  • Typ balení:
    Bottle
  • Sekvence:
    2725

Specifications