Mitomycin
2590, Bottle
Catalog # APOSBIM0133-250MG
New Product
Supplier: Apollo Scientific
Jump to:
Product Details & Documents
Mitomycin C is a HIGHLY TOXIC antibiotic with antineoplastic properties.
Synonyms: Ametycin
- CAS Number:50-07-7
- Formula:C₁₅H₁₈N₄O₅
- Melting Pt:360 °C
- Storage Temperature:Refrigerator
- EINECS:200-008-6
- UN:3462
- ADR:6.1,II
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Typ balení:Bottle
- Sekvence:2590
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



