Orders LinkContactUsLinkComponent
Picolinamide ≥98%
2070, Bottle
  APOSBUP18994-1ML
New Product
Picolinamide ≥98%
  APOSBUP18994-1ML
New Product
 :  1452-77-3
Picolinamide ≥98%
2070, Bottle
 :  1452-77-3
1 070,00 Kč
1 * 1 mL
Usage: Biochemical/ life science applications.
 :  2-Pyridinecarboxamide, Pyridine-2-carboxamide
  •  :1452-77-3
  •  :C₆H₆N₂O
  •  :122,12 g/mol
  • Typ balení:
    Bottle
  • Sekvence:
    2070