Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Ethionamide ≥98%

Ethionamide ≥98%

New Product
Dodavatel:   Apollo Scientific
Ethionamide ≥98%
Dodavatel:  Apollo Scientific
New Product
Certifikáty
Ethionamide ≥98%

3 Dostupných variant níže

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyma:  2-Ethylpyridine-4-carbothioamide, 2-Ethylthioisonicotinamide
  • Číslo CAS:536-33-4
  • Vzorec:C₈H₁₀N₂S
  • Molecular Weight:166,24 g/mol
Product Family Options

Informace o produktu

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.