Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
Obefazimod ≥98%

Obefazimod ≥98%

New Product
Supplier:   Apollo Scientific
Obefazimod ≥98%
Supplier:  Apollo Scientific
New Product
Certificates
Obefazimod ≥98%

4 Options Available Below

Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  Obefazimod
  • CAS Number:1258453-75-6
  • Formula:C₁₆H₁₀ClF₃N₂O
  • Molecular Weight:338,71 g/mol
Product Family Options

Product Information

  • Sekvence
    Typ balení
    Availability
    Price
  • 2315
    Bottle
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 7 100,00 Kč
    7 100,00 Kč
    1 * 25 mg
       7 100,00 Kč

    Specifications:
    New Product
    New Product
  • 2370
    Bottle
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - 11 440,00 Kč
    11 440,00 Kč
    1 * 50 mg
       11 440,00 Kč

    Specifications:
    New Product
    New Product
  • 2440
    Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 16 600,00 Kč
    16 600,00 Kč
    1 * 100 mg
       16 600,00 Kč

    Specifications:
    New Product
    New Product
  • 2530
    Bottle
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 23 370,00 Kč
    23 370,00 Kč
    1 * 200 mg
       23 370,00 Kč

    Specifications:
    New Product
    New Product