Orders LinkContactUsLinkComponent
CLASSIQSwabs™ Regular Polyester Swabs
CLASSIQSwabs™ Regular Polyester Swabs
Single polyester regular swab, with 100 mm breakpoint, Polyester, +
Catalog # COPA1U054S01
New Product
Supplier:   Copan
CLASSIQSwabs™ Regular Polyester Swabs
Catalog # COPA1U054S01
Supplier:  Copan
New Product
CLASSIQSwabs™ Regular Polyester Swabs
Single polyester regular swab, with 100 mm breakpoint, Polyester, +
Price
4 730,00 Kč
Log in for availability
1 * 1.000 KS
Product Details & Documents
  • Traditional polyester nasal
    Individually packaged
    Sterile
 : 100 swabs per box, 1000 swabs per case.
Specifications
  • Sterilní:
    +
  • Popis:
    Single polyester regular swab, with 100 mm breakpoint
  • Materiál:
    Polyester