Optical Microscope
optical, Min.: 10 unit, Max.: 50 unit, LED illumination
Katalogové číslo IDCPAF7115MZTW
New Product
Dodavatel: Dino-Lite / IDCP B.V.
Přejít na:
Product Details & Documents
The 5MP Edge AM7115MZTW - Edge provides macro-level imaging with zoomin capabilities. This microscope includes a polariser to reduce reflections and features FLC for adjustable illumination. AF7115MZTW is compatible with the optional WF-20 Wi-Fi streamer.
5 Megapixel sensor
Macro zoom optics
Adjustable polariser
Robust metal housing
Flexible LED control (FLC)
High optical resolution
Adaptable interface
Specifikace
- Zvětšení:Min.: 10 unit, Max.: 50 unit
- Osvětlení:LED illumination
- Typ:optical
- Hmotnost:97 g
- Cat. No.:IDCPAF7115MZTW
- Kamera:Digital camera
- Kontrastní technika:Polarizace