Combination Blocks
New Product
Supplier: VELP SCIENTIFIC
Jump to:
About this item
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- SekvencePopisRozměry jamkyUrčeno proPoč. otvorůHloubkaAvailabilityPrice
- 3MCT-W21-COMBO blockØ 7,9/11,1/11,5 mm0,5/1,5/2,0 ml microcentrifuge tubes6/10/527,6/39,1/38,1 mmLog in for availability1 * 1 KS3 210,00 KčSpecifications:New ProductNew Product
- 1STT-W14-COMBO blockØ 8,3/13,9/26,2 mm6/12/13/25 mm round-bottom tubes6/5/348,4 mmLog in for availability1 * 1 KS3 210,00 KčSpecifications:New ProductNew Product
- 2CCT-W09-COMBO blockØ 11,1/17,1/29 mm1,5/15/50 ml conical centrifuge4/3/239,1/44,5/47,6 mmLog in for availability1 * 1 KS3 210,00 KčSpecifications:New ProductNew Product