Explorer Plus™ Precision Balances
O tomto produktu
Welcome to the future of laboratory weighing with the OHAUS Explorer Plus™ EXP series. This new generation of laboratory balances is designed to enhance your workflow with unparalleled user experience, outstanding weighing performance, advanced communication capabilities, and an eco-friendly design.
7" touchscreen display; vibrant colours, an easy-to-clean glass panel, and chemical resistance ensure a clear and durable interface
- Detachable terminal and base; designed for flexibility, and suitable for small laboratory cabinets or fume hoods
- Effortless setup and monitoring; motorised levelling system with a digital-level bubble, that ensures a smooth and straightforward setup
- Intuitive sample handling and reliable status indicators; features an intuitive sample spotlight that provides clear visibility, ensuring precise and hassle-free sample handling
- Customised terminal and draft shield status lights (on 1mg models); provide clear and immediate feedback to underload or overload conditions
- Enhanced anti-pinch automatic draft shield door; safety protection with IR sensor operation for easy sample access and preventing hand injuries (on 1mg models)
- Half-open design - tailored for powder samples that require a longer time to stabilise with airflow (on 1mg models)
- Internal Calibration Systems; Automatic, user-triggered, and span calibration options ensure precise measurements
Engineered for accuracy, designed for demanding applications: Standard Square Weighing Pan - designed for laboratory efficiency and specifically designed to assist in sample preparation. Sample Libraries shared among applications: Apply to 15 weighing applications and 3000 sample libraries for Explorer Plus EXP.
Removable Draft shield (on 1 mg models): For easy cleaning and long-term care and specific large volume weighing applications. Digital High-Speed Load Cell; The digital weighing components (DWC) offer the advantage of creating self-contained load cell modules. Rapid and precise measurements with this advanced technology ensures weighing processes are both swift and highly accurate. Elevated Digital Load Cell Platform: Equipped with three ambient sensors to measure key load cell factors - temperature, humidity, and air pressure. Comprehensive monitoring ensures optimal weighing conditions, enhancing the accuracy and reliability of the measurements.
Anti-Static Draft shield (on 1 mg models): Reduces static electricity around the balance for consistent results. Built-In Ioniser in Draft shield models: Removes the static charge from the sample surface and neutralizes the weighing chamber for a stable environment. Routine Repeatability Test: Ensures consistent long-term accuracy and reliability. Ideal for critical applications such as weighing in regulated environments, where meeting strict uncertainty requirements is essential (e.g., pharmacopeia mandates uncertainty ≤0.10%).
Supports compliance for data traceability: Equipped with multiple USB ports; offering unparalleled flexibility, the direct to Excel feature provides a basic yet highly user-friendly solution for data transfer. Human Interface Device allows connections to computers without the need for drivers and easy integration with LIMS and MES systems. Time Synchronisation - Network Time Protocol (NTP) ensures accurate timekeeping. System Log: All the changes made on balance setting will keep records in system log file. These system log files can be reviewed and exported as a PDF to a USB flash drive. Direct Data Transfer; Direct export to Excel and saving data to USB in CSV and PDF formats. Label Printing Function: two preset templates and 10 customisable templates for semi-finished product samples transfer management. When the Electronic Signature feature is enabled, the User ID will be used in the printout field when the User ID content option is selected.
Eco-Friendly Design: 65% reduced power consumption in Sleep Mode. Reduced shipping weight; 36% weight reduction for non-draft shield models, 64% weight reduction for draft shield models. Recycled materials, up to 58% of total components. Aluminum construction: Replaces most plastic materials for durability and sustainability. Dematerialisation: Reduces the number of components to improve production efficiency. ACT Label Certification: 11 models are certified with the ACT Label, highlighting their environmental impact.
Certifications: OIML R76, EN 45501
IEC/EN 61010-1; CAN/CSA C22.2 61010-1
UL 61010-1
IEC/EN 61326-1 Class B, Basic Environments
FCC Part 15 Class A
Canada ICES-003 Class A.
Delivery information: Supplied with AC adapter and a plastic in use protective cover.
24 Dostupných variant níže
Product Details & Documents
Welcome to the future of laboratory weighing with the OHAUS Explorer Plus™ EXP series. This new generation of laboratory balances is designed to enhance your workflow with unparalleled user experience, outstanding weighing performance, advanced communication capabilities, and an eco-friendly design.
7" touchscreen display; vibrant colours, an easy-to-clean glass panel, and chemical resistance ensure a clear and durable interface
- Detachable terminal and base; designed for flexibility, and suitable for small laboratory cabinets or fume hoods
- Effortless setup and monitoring; motorised levelling system with a digital-level bubble, that ensures a smooth and straightforward setup
- Intuitive sample handling and reliable status indicators; features an intuitive sample spotlight that provides clear visibility, ensuring precise and hassle-free sample handling
- Customised terminal and draft shield status lights (on 1mg models); provide clear and immediate feedback to underload or overload conditions
- Enhanced anti-pinch automatic draft shield door; safety protection with IR sensor operation for easy sample access and preventing hand injuries (on 1mg models)
- Half-open design - tailored for powder samples that require a longer time to stabilise with airflow (on 1mg models)
- Internal Calibration Systems; Automatic, user-triggered, and span calibration options ensure precise measurements
Engineered for accuracy, designed for demanding applications: Standard Square Weighing Pan - designed for laboratory efficiency and specifically designed to assist in sample preparation. Sample Libraries shared among applications: Apply to 15 weighing applications and 3000 sample libraries for Explorer Plus EXP.
Removable Draft shield (on 1 mg models): For easy cleaning and long-term care and specific large volume weighing applications. Digital High-Speed Load Cell; The digital weighing components (DWC) offer the advantage of creating self-contained load cell modules. Rapid and precise measurements with this advanced technology ensures weighing processes are both swift and highly accurate. Elevated Digital Load Cell Platform: Equipped with three ambient sensors to measure key load cell factors - temperature, humidity, and air pressure. Comprehensive monitoring ensures optimal weighing conditions, enhancing the accuracy and reliability of the measurements.
Anti-Static Draft shield (on 1 mg models): Reduces static electricity around the balance for consistent results. Built-In Ioniser in Draft shield models: Removes the static charge from the sample surface and neutralizes the weighing chamber for a stable environment. Routine Repeatability Test: Ensures consistent long-term accuracy and reliability. Ideal for critical applications such as weighing in regulated environments, where meeting strict uncertainty requirements is essential (e.g., pharmacopeia mandates uncertainty ≤0.10%).
Supports compliance for data traceability: Equipped with multiple USB ports; offering unparalleled flexibility, the direct to Excel feature provides a basic yet highly user-friendly solution for data transfer. Human Interface Device allows connections to computers without the need for drivers and easy integration with LIMS and MES systems. Time Synchronisation - Network Time Protocol (NTP) ensures accurate timekeeping. System Log: All the changes made on balance setting will keep records in system log file. These system log files can be reviewed and exported as a PDF to a USB flash drive. Direct Data Transfer; Direct export to Excel and saving data to USB in CSV and PDF formats. Label Printing Function: two preset templates and 10 customisable templates for semi-finished product samples transfer management. When the Electronic Signature feature is enabled, the User ID will be used in the printout field when the User ID content option is selected.
Eco-Friendly Design: 65% reduced power consumption in Sleep Mode. Reduced shipping weight; 36% weight reduction for non-draft shield models, 64% weight reduction for draft shield models. Recycled materials, up to 58% of total components. Aluminum construction: Replaces most plastic materials for durability and sustainability. Dematerialisation: Reduces the number of components to improve production efficiency. ACT Label Certification: 11 models are certified with the ACT Label, highlighting their environmental impact.
Certifications: OIML R76, EN 45501
IEC/EN 61010-1; CAN/CSA C22.2 61010-1
UL 61010-1
IEC/EN 61326-1 Class B, Basic Environments
FCC Part 15 Class A
Canada ICES-003 Class A.
Delivery information: Supplied with AC adapter and a plastic in use protective cover.
Specifications for Product Family
Specifikace
- Catalog No:
- OHAU31059084
- OHAU31059085
- OHAU31059086
- OHAU31059087
- OHAU31059099
- OHAU31059100
- OHAU31059101
- OHAU31059102
- OHAU31059103
- OHAU31059104
- OHAU31059105
- OHAU31059106
- OHAU31059095
- OHAU31059096
- OHAU31059097
- OHAU31059098
- OHAU31059107
- OHAU31059108
- OHAU31059109
- OHAU31059110
- OHAU31059111
- OHAU31059112
- OHAU31059113
- OHAU31059114
- Model:
- EXP223/AD
- EXP423/AD
- EXP623/AD
- EXP1203/AD
- EXP2202
- EXP4202
- EXP6202
- EXP8202
- EXP12202
- EXP6201
- EXP8201
- EXP10201
- EXP223M/AD
- EXP423M/AD
- EXP623M/AD
- EXP1203M/AD
- EXP2202M
- EXP4202M
- EXP6202M
- EXP8202M
- EXP12202M
- EXP6201M
- EXP8201M
- EXP10201M
- Rozsah provozních teplot:
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- 10…30 °C
- Typ přípojky:
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Supplied with AC adapter
- Komunikační rozhraní:
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Ethernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)
- Napětí:
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
- 100 - 240 V
Specifikace
Product Family Options
1 mg Precision balances (with draft shield)
Informace o produktu
- SekvenceModelHmotnostní rozsahČitelnostMiska/velikost plošinyOpakovatelnostLinearitaDoba stabilizaceRozměryDostupnostPrice
- 0EXP223/AD220 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS94 200,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059084ModelEXP223/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 2EXP423/AD420 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS110 190,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059085ModelEXP423/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 3EXP623/AD620 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS134 650,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059086ModelEXP623/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 4EXP1203/AD1200 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS157 900,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059087ModelEXP1203/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product
0,01 g Precision balances
Informace o produktu
- SekvenceModelHmotnostní rozsahČitelnostMiska/velikost plošinyOpakovatelnostLinearitaDoba stabilizaceRozměryDostupnostPrice
- 5EXP22022200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS99 170,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059099ModelEXP2202Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 6EXP42024200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS113 950,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059100ModelEXP4202Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 7EXP62026200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS129 270,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059101ModelEXP6202Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 8EXP82028200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS141 230,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059102ModelEXP8202Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 9EXP1220212200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS153 330,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059103ModelEXP12202Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product
0,1 g Precision balances
Informace o produktu
- SekvenceModelHmotnostní rozsahČitelnostMiska/velikost plošinyOpakovatelnostLinearitaDoba stabilizaceRozměryDostupnostPrice
- 10EXP62016200 g0,1 g201×178 mm0,1 g0,2 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS98 100,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059104ModelEXP6201Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 11EXP82018200 g0,1 g201×178 mm0,1 g0,2 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS107 910,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059105ModelEXP8201Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 12EXP1020110200 g0,1 g201×178 mm0,1 g0,2 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS117 720,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059106ModelEXP10201Rozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product
1 mg Approved precision balances (with draft shield)
Informace o produktu
- SekvenceModelHmotnostní rozsahČitelnostMiska/velikost plošinyOpakovatelnostLinearitaDoba stabilizaceRozměryDostupnostPrice
- 13EXP223M/AD220 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS102 940,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059095ModelEXP223M/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 14EXP423M/AD420 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS118 930,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059096ModelEXP423M/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 15EXP623M/AD620 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS143 380,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059097ModelEXP623M/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 16EXP1203M/AD1200 g1 mg130×130 mm1 mg2 mg1,5 s372×408×229 mmPřihlásit se pro dostupnost1 * 1 KS166 630,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059098ModelEXP1203M/ADRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product
0,01 g Approved precision balances
Informace o produktu
- SekvenceModelHmotnostní rozsahČitelnostMiska/velikost plošinyOpakovatelnostLinearitaDoba stabilizaceRozměryDostupnostPrice
- 17EXP2202M2200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS107 910,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059107ModelEXP2202MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 18EXP4202M4200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS122 690,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059108ModelEXP4202MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 19EXP6202M6200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS138 010,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059109ModelEXP6202MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 20EXP8202M8200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS149 970,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059110ModelEXP8202MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 21EXP12202M12200 g0,01 g201×178 mm0,01 g0,02 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS162 060,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059111ModelEXP12202MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product
0,1 g Approved precision balances
Informace o produktu
- SekvenceModelHmotnostní rozsahČitelnostMiska/velikost plošinyOpakovatelnostLinearitaDoba stabilizaceRozměryDostupnostPrice
- 22EXP6201M6200 g0,1 g201×178 mm0,1 g0,2 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS106 830,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059112ModelEXP6201MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 23EXP8201M8200 g0,1 g201×178 mm0,1 g0,2 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS116 640,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059113ModelEXP8201MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product - 24EXP10201M10200 g0,1 g201×178 mm0,1 g0,2 g1 s97×408×229 mmPřihlásit se pro dostupnost1 * 1 KS126 450,00 KčSpecifikace:New Product
More
Specifications
Cat. No.OHAU31059114ModelEXP10201MRozsah provozních teplot10…30 °CTyp přípojkySupplied with AC adapterKomunikační rozhraníEthernet TCP/IP;RS232;USB device;USB host x 2;WiFi/Bluetooth (available as an accessory)Napětí100 - 240 VNew Product