QUANTOFIX® Test Strips, Chlorine dioxide
O tomto produktu
Semi-quantitative test strips for the determination of chlorine dioxide.
Chlorine dioxide - 0, 10, 25, 50, 100, 250, 500, 1000, 1500, 2000, 2500, 3000 mg/L
- Chlorine dioxide - Yellow → dark green (field 1), colourless → brown-black (field 2)
Ideal for rapid and easy analysis of chlorine dioxide directly on-site. No maintenance or calibration is required.
2 Dostupných variant níže
Product Details & Documents
Semi-quantitative test strips for the determination of chlorine dioxide.
Chlorine dioxide - 0, 10, 25, 50, 100, 250, 500, 1000, 1500, 2000, 2500, 3000 mg/L
- Chlorine dioxide - Yellow → dark green (field 1), colourless → brown-black (field 2)
Ideal for rapid and easy analysis of chlorine dioxide directly on-site. No maintenance or calibration is required.
Specifications for Product Family
Specifikace
Specifikace
- Catalog No:
- MANA91355
- MANA91357
- Metoda:
- Chlorine dioxide - Redox reaction (colorless → pink)
- Chlorine dioxide - Redox reaction (Yellow → dark green (field 1), colorless → brown-black (field 2))
- Parametry:
- All reagents included
- All reagents included
- Shoda:
- No hazardous material
- No hazardous material
- Obsah:
- 50 test strips in a tube, instruction leaflet
- 100 test strips in a tube, instruction leaflet
- Přístroj:
- No
- No
- Plošina:
- Semi-quantitative test strips
- Semi-quantitative test strips
- Životnost police (měsíce):
- 24 (from production)
- 30 (from production)
- Hmotnost:
- 35 g
- 42.8 g
Specifikace
Product Family Options
Informace o produktu
- SekvenceRozsahChemická metodaRozměryTestů v kituDostupnostPrice
- MANA91355QUANTOFIX Chlorine dioxide 15QUANTOFIX Chlorine dioxide 1510.25 • 0.5 • 1 • 3 • 5 • 10 • 15 - mg/LColour reaction with a triazine derivate40×40×120 mm (package)50 test(s)Přihlásit se pro dostupnost1 * 50 Tests679,00 KčSpecifikace:
More
Specifications
Cat. No.MANA91355MetodaChlorine dioxide - Redox reaction (colorless → pink)ParametryAll reagents includedShodaNo hazardous materialObsah50 test strips in a tube, instruction leafletPřístrojNoPlošinaSemi-quantitative test stripsŽivotnost police (měsíce)24 (from production)Hmotnost35 g - MANA91357QUANTOFIX Dioxyde de chlore 3000QUANTOFIX Dioxyde de chlore 300020 • 10 • 25 • 50 • 100 • 250 • 500 • 1000 • 1500 • 2000 • 2500 • 3000 - mg/LRedox reaction (potassium iodide / redox indicator)40×40×120 mm (package)100 test(s)Přihlásit se pro dostupnost1 * 100 Tests963,00 KčSpecifikace:
More
Specifications
Cat. No.MANA91357MetodaChlorine dioxide - Redox reaction (Yellow → dark green (field 1), colorless → brown-black (field 2))ParametryAll reagents includedShodaNo hazardous materialObsah100 test strips in a tube, instruction leafletPřístrojNoPlošinaSemi-quantitative test stripsŽivotnost police (měsíce)30 (from production)Hmotnost42.8 g